Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CSTA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005547994.2 | XP_005548051.1 | 2 - 3 | 291 | 168 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Probe spans exons |