Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BTBD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005568343.2 | XP_005568400.1 | 2 - 3 | 454 | 88 | |
XM_005568345.2 | XP_005568402.1 | 2 - 3 | 8777 | 88 | ||
XM_015457780.1 | XP_015313266.1 | 2 - 3 | 282 | 88 | ||
XM_015457783.1 | XP_015313269.1 | 2 - 3 | 279 | 88 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Probe spans exons |