Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PYGO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015452865.1 | XP_015308351.1 | 2 - 3 | 401 | 87 | |
XM_015452866.1 | XP_015308352.1 | 2 - 3 | 514 | 87 | ||
XM_015452867.1 | XP_015308353.1 | 2 - 3 | 401 | 87 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Probe spans exons |