Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CFAP53 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005586703.2 | XP_005586760.1 | 6 - 7 | 1468 | 80 | |
XM_015439966.1 | XP_015295452.1 | 6 - 7 | 1468 | 80 | ||
XM_015439967.1 | XP_015295453.1 | 6 - 7 | 1468 | 80 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Probe spans exons |