Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZDHHC23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005548106.2 | XP_005548163.1 | 3 - 4 | 1091 | 100 | |
XM_015446400.1 | XP_015301886.1 | 3 - 4 | 1188 | 100 | ||
XM_015446401.1 | XP_015301887.1 | 3 - 4 | 1226 | 100 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Probe spans exons |