Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAAH2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005593740.2 | XP_005593797.1 | 3 - 4 | 657 | 104 | |
XM_005593742.2 | XP_005593799.1 | 3 - 4 | 310 | 104 | ||
XM_015443530.1 | XP_015299016.1 | 4 - 5 | 779 | 104 | ||
GenBank mRNA | EU285561.1 | - | 3 - 4 | 414 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Probe spans exons |