Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TAFA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001285077.1 | NP_001272006.1 | 2 - 3 | 559 | 68 | |
XM_005547566.2 | XP_005547623.1 | 2 - 3 | 384 | 68 | ||
GenBank mRNA | AB172528.1 | - | 2 - 3 | 559 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Probe spans exons |