Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C3AR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005570035.2 | XP_005570092.1 | 1 - 2 | 285 | 73 |
Set Membership: |
Amplicon length between 71 and 85 G-protein coupled receptor Inventoried Probe spans exons |