Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPRY3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015443910.1 | XP_015299396.1 | 1 - 2 | 401 | 95 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Probe spans exons |