Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PROX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005561780.2 | XP_005561837.1 | 3 - 4 | 1594 | 77 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Probe spans exons |