Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EEF1AKMT3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005571389.2 | XP_005571446.1 | 2 - 3 | 437 | 94 | |
XM_015431130.1 | XP_015286616.1 | 2 - 3 | 207 | 94 | ||
XM_015431131.1 | XP_015286617.1 | 2 - 3 | 303 | 94 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Probe spans exons |