Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARMCX5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005594203.1 | XP_005594260.1 | 6 | 2492 | 101 | |
XM_005594204.1 | XP_005594261.1 | 6 | 2486 | 101 | ||
XM_005594205.1 | XP_005594262.1 | 5 | 2373 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Both primers and probe map within a single exon |