Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IGSF6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001283642.1 | NP_001270571.1 | 5 - 6 | 694 | 97 | |
XM_005591446.2 | XP_005591503.1 | 5 - 6 | 836 | 97 | ||
GenBank mRNA | AB168410.1 | - | 5 - 6 | 694 | 97 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Probe spans exons |