Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SRPK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001285355.1 | NP_001272284.1 | 11 - 12 | 1738 | 72 | |
XM_015447283.1 | XP_015302769.1 | 10 - 11 | 1682 | 72 | ||
XM_015447284.1 | XP_015302770.1 | 10 - 11 | 2924 | 72 | ||
XM_015447285.1 | XP_015302771.1 | 10 - 11 | 2045 | 72 | ||
XM_015447286.1 | XP_015302772.1 | 11 - 12 | 1746 | 72 | ||
XM_015447287.1 | XP_015302773.1 | 10 - 11 | 1683 | 72 | ||
XM_015447288.1 | XP_015302774.1 | 10 - 11 | 1681 | 72 | ||
XM_015447289.1 | XP_015302775.1 | 10 - 11 | 1682 | 72 | ||
XM_015447290.1 | XP_015302776.1 | 10 - 11 | 1744 | 72 | ||
XM_015447292.1 | XP_015302778.1 | 10 - 11 | 1682 | 72 | ||
GenBank mRNA | AB168194.1 | - | 11 - 12 | 1738 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Probe spans exons |