Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRRD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001283368.1 | NP_001270297.1 | 3 - 4 | 2318 | 85 | |
XM_015447256.1 | XP_015302742.1 | 4 - 5 | 1055 | 85 | ||
GenBank mRNA | AB168315.1 | - | 2 - 3 | 1436 | 85 | |
AB169234.1 | - | 3 - 4 | 2318 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Probe spans exons |