Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BMF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005559153.2 | XP_005559210.1 | 4 - 5 | 635 | 98 | |
XM_015452634.1 | XP_015308120.1 | 3 - 4 | 901 | 98 | ||
XM_015452635.1 | XP_015308121.1 | 3 - 4 | 554 | 98 | ||
XM_015452636.1 | XP_015308122.1 | 2 - 3 | 1614 | 98 | ||
XM_015452637.1 | XP_015308123.1 | 2 - 3 | 1090 | 98 | ||
XM_015452638.1 | XP_015308124.1 | 2 - 3 | 1518 | 98 |