Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM9C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005592975.2 | XP_005593032.1 | 6 - 7 | 1182 | 81 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Probe spans exons |