Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VAX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015456759.1 | XP_015312245.1 | 1 - 2 | 1685 | 79 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Probe spans exons |