Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCIN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005581326.2 | XP_005581383.1 | 1 | 1571 | 90 | |
XM_005581327.2 | XP_005581384.1 | 1 | 1578 | 90 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Both primers and probe map within a single exon |