Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TREML4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005595974.2 | XP_005596031.2 | 2 - 3 | 480 | 118 | |
XM_015449422.1 | XP_015304908.1 | 2 - 3 | 480 | 118 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Probe spans exons |