Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SIGLEC15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005586809.2 | XP_005586866.1 | 1 - 2 | 465 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Recommended Assay Probe spans exons |