Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADGRE3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015440203.1 | XP_015295689.1 | 3 - 4 | 695 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 G-protein coupled receptor Probe spans exons |