Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TTLL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005551436.2 | XP_005551493.1 | 1 - 2 | 423 | 91 | |
GenBank mRNA | AB169207.1 | - | 1 - 2 | 114 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Probe spans exons |