Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EMILIN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005569018.2 | XP_005569075.1 | 2 - 3 | 304 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Recommended Assay Probe spans exons |