Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FBXO39 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005582678.2 | XP_005582735.1 | 3 - 4 | 1388 | 85 | |
XM_005582680.2 | XP_005582737.1 | 3 - 4 | 1369 | 85 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Probe spans exons |