Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SGK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005551891.2 | XP_005551948.2 | 1 - 2 | 447 | 73 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Probe spans exons |