Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DPRX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005590262.2 | XP_005590319.1 | 1 - 2 | 325 | 75 | |
XM_005590264.2 | XP_005590321.1 | 4 - 5 | 465 | 75 | ||
XM_005590265.2 | XP_005590322.1 | 3 - 4 | 338 | 75 | ||
XM_015442100.1 | XP_015297586.1 | 3 - 4 | 779 | 75 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Probe spans exons |