Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRRT4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015447359.1 | XP_015302845.1 | 2 - 3 | 817 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Probe spans exons |