Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IQCA1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005551202.2 | XP_005551259.2 | 15 - 16 | 1936 | 99 | |
XM_015448298.1 | XP_015303784.1 | 15 - 16 | 1937 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Probe spans exons |