Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RUNDC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005584317.2 | XP_005584374.1 | 4 - 5 | 925 | 67 | |
XM_005584318.2 | XP_005584375.1 | 3 - 4 | 919 | 67 | ||
XM_005584319.2 | XP_005584376.1 | 4 - 5 | 925 | 67 | ||
XM_005584320.2 | XP_005584377.1 | 3 - 4 | 919 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Probe spans exons |