Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC102144954 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005541004.2 | XP_005541061.1 | 1 | 528 | 122 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Both primers and probe map within a single exon |