Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCNYL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001287626.1 | NP_001274555.1 | 6 - 7 | 849 | 119 | |
XM_005564999.2 | XP_005565056.1 | 6 - 7 | 856 | 119 | ||
GenBank mRNA | AB168587.1 | - | 6 - 7 | 849 | 119 |
Set Membership: |
Amplicon length greater than or equal to 101 Probe spans exons |