Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bmp6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007556.2 | NP_031582.1 | 3 - 4 | 1213 | 88 | |
GenBank mRNA | AK041210.1 | - | 1571 | 88 | ||
AK135770.1 | - | 3 - 4 | 846 | 88 | ||
BC138593.1 | - | 3 - 4 | 1133 | 8861613 | 88 | |
BC138595.1 | - | 3 - 4 | 1133 | 8861615 | 88 | |
J04566.1 | - | 3 - 4 | 795 | 88 | ||
X80992.1 | - | 3 - 4 | 1212 | 88 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |