Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccnf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007634.4 | NP_031660.3 | 5 - 6 | 643 | 63 | |
GenBank mRNA | AK030298.1 | - | 582 | 63 | ||
AK031613.1 | - | 5 - 6 | 583 | 63 | ||
AK032961.1 | - | 5 - 6 | 569 | 63 | ||
AK082485.1 | - | 5 - 6 | 611 | 63 | ||
AK085183.1 | - | 5 - 6 | 580 | 63 | ||
AK159594.1 | - | 5 - 6 | 644 | 63 | ||
AK162447.1 | - | 5 - 6 | 583 | 63 | ||
AK169263.1 | - | 5 - 6 | 594 | 63 | ||
BC021658.1 | - | 5 - 6 | 563 | 63 | ||
BC037662.1 | - | 5 - 6 | 595 | 5353169 | 63 | |
U20612.1 | - | 5 - 6 | 581 | 63 | ||
U20636.1 | - | 5 - 6 | 698 | 63 | ||
Z47766.1 | - | 5 - 6 | 583 | 63 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |