Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1500016550.mm.1 |
NM_007692.6.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500016550.mm.1 |
NM_007692.6 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500016550.mm.1 |
NR_037153.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500016550.mm.1 |
NR_037153.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500016550.mm.1 |
NR_037154.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500016550.mm.1 |
NR_037154.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500016550.mm.1 |
TC1500002351.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1500016556.mm.1 |
NM_007692.6.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500016556.mm.1 |
NM_007692.6 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500016556.mm.1 |
NR_037153.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500016556.mm.1 |
NR_037153.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500016556.mm.1 |
NR_037154.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500016556.mm.1 |
NR_037154.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500016556.mm.1 |
TC1500002351.mm.1 |
MTA TC |