Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ebf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001290709.1 | NP_001277638.1 | 13 - 14 | 1450 | 63 | |
NM_001290711.1 | NP_001277640.1 | 13 - 14 | 1566 | 63 | ||
NM_007897.3 | NP_031923.1 | 13 - 14 | 1447 | 63 | ||
GenBank mRNA | AK031239.1 | - | 13 - 14 | 1445 | 63 | |
AK035781.1 | - | 13 - 14 | 1330 | 63 | ||
AK137389.1 | - | 13 - 14 | 1488 | 63 | ||
AK141175.1 | - | 13 - 14 | 1450 | 63 | ||
BC139362.1 | - | 13 - 14 | 1408 | 8862384 | 63 | |
BC139364.1 | - | 13 - 14 | 1408 | 8862386 | 63 | |
BC139765.1 | - | 6 - 7 | 899 | 63 | ||
L12147.1 | - | 13 - 14 | 1413 | 63 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |