Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eef2k | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001267710.1 | NP_001254639.1 | 5 - 6 | 709 | 64 | |
NM_001267711.1 | NP_001254640.1 | 5 - 6 | 704 | 64 | ||
NM_007908.4 | NP_031934.1 | 5 - 6 | 793 | 64 | ||
GenBank mRNA | AK080915.1 | - | 5 - 6 | 710 | 64 | |
AK135891.1 | - | 3 - 4 | 135 | 64 | ||
AK136015.1 | - | 5 - 6 | 705 | 64 | ||
BC003433.1 | - | 5 - 6 | 741 | 2646503 | 64 | |
BC055361.1 | - | 5 - 6 | 704 | 30046537 | 64 | |
U93848.1 | - | 4 - 5 | 447 | 64 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Protein Kinase Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |