Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc6a9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008135.4 | NP_032161.2 | 7 - 8 | 1167 | 78 | |
GenBank mRNA | AK014572.1 | - | 1011 | 78 | ||
BC021828.1 | - | 7 - 8 | 990 | 4194951 | 78 | |
X67056.1 | - | 7 - 8 | 1100 | 78 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Transporter Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |