Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Grin2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008171.3 | NP_032197.3 | 12 - 13 | 2932 | 71 | |
GenBank mRNA | D10651.2 | - | 9 - 10 | 2169 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Ion Channel Mus musculus Probe spans exons |