Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cfb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001142706.1 | NP_001136178.1 | 8 - 9 | 1567 | 71 | |
NM_008198.2 | NP_032224.2 | 8 - 9 | 1567 | 71 | ||
GenBank mRNA | AK098094.1 | - | 8 - 9 | 1227 | 71 | |
AK149531.1 | - | 8 - 9 | 1200 | 71 | ||
AK149573.1 | - | 8 - 9 | 1226 | 71 | ||
AK151220.1 | - | 2 - 3 | 186 | 71 | ||
AK151618.1 | - | 2 - 3 | 186 | 71 | ||
AK151974.1 | - | 2 - 3 | 186 | 71 | ||
BC005451.1 | - | 8 - 9 | 1201 | 3591669 | 71 | |
K01496.1 | - | 3 - 4 | 307 | 71 | ||
K01497.1 | - | 2 - 3 | 166 | 71 | ||
M57890.1 | - | 8 - 9 | 1270 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protease Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |