Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Il13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008355.3 | NP_032381.1 | 2 - 3 | 266 | 74 | |
GenBank mRNA | M23504.1 | - | 2 - 3 | 267 | 74 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Apoptosis Detect Genomic DNA Mus musculus Inventoried Probe spans exons |