Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Itih3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008407.2 | NP_032433.2 | 9 - 10 | 1118 | 67 | |
GenBank mRNA | BC015276.1 | - | 9 - 10 | 1076 | 4212290 | 67 |
X70393.1 | - | 9 - 10 | 1075 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |