Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmprss15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008941.3 | NP_032967.1 | 18 - 19 | 2090 | 102 | |
NM_178855.4 | NP_849186.2 | 17 - 18 | 2045 | 102 | ||
GenBank mRNA | AK033582.1 | - | 17 - 18 | 2047 | 102 | |
AK038356.1 | - | 9 - 10 | 1039 | 102 | ||
BC117917.1 | - | 18 - 19 | 2089 | 40097367 | 102 | |
BC117918.1 | - | 18 - 19 | 2089 | 40097370 | 102 | |
U73378.1 | - | 18 - 19 | 2085 | 102 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |