Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rps6ka1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001285505.1 | NP_001272434.1 | 7 - 8 | 739 | 65 | |
NM_001285506.1 | NP_001272435.1 | 6 - 7 | 590 | 65 | ||
NM_009097.5 | NP_033123.2 | 7 - 8 | 739 | 65 | ||
GenBank mRNA | AK132856.1 | - | 5 - 6 | 776 | 65 | |
AK148208.1 | - | 7 - 8 | 732 | 65 | ||
AK167790.1 | - | 6 - 7 | 577 | 65 | ||
BC049076.1 | - | 7 - 8 | 690 | 65 | ||
BC094470.1 | - | 7 - 8 | 692 | 5098650 | 65 | |
M28489.1 | - | 7 - 8 | 725 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Protein Kinase Mammalian Gene Collection Mus musculus Probe spans exons |