Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc4a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001253892.1 | NP_001240821.1 | 7 - 8 | 1204 | 89 | |
NM_009207.3 | NP_033233.2 | 8 - 9 | 1400 | 89 | ||
GenBank mRNA | AK145930.1 | - | 8 - 9 | 1325 | 89 | |
AK147858.1 | - | 8 - 9 | 1356 | 89 | ||
BC054102.1 | - | 7 - 8 | 1209 | 30017310 | 89 | |
J04036.1 | - | 8 - 9 | 1315 | 89 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Transporter Mammalian Gene Collection Mus musculus Probe spans exons |