Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chrng | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009604.3 | NP_033734.3 | 10 - 11 | 1265 | 61 | |
GenBank mRNA | AK047974.1 | - | 10 - 11 | 1266 | 61 | |
M30514.1 | - | 10 - 11 | 1306 | 61 | ||
X03818.1 | - | 10 - 11 | 1262 | 61 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Ion Channel Detect Genomic DNA Mus musculus Probe spans exons |