Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atp7a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109757.2 | NP_001103227.1 | 18 - 19 | 3812 | 74 | |
NM_009726.5 | NP_033856.3 | 18 - 19 | 3809 | 74 | ||
GenBank mRNA | AB007134.1 | - | 17 - 18 | 3648 | 74 | |
BC141395.1 | - | 18 - 19 | 3690 | 9056008 | 74 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Ion Channel Transporter Mammalian Gene Collection Mus musculus Probe spans exons |