Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dok1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010070.4 | NP_034200.4 | 1 - 2 | 100 | 79 | |
GenBank mRNA | AK136382.1 | - | 1 - 2 | 101 | 79 | |
BC013066.1 | - | 1 - 2 | 94 | 3498170 | 79 | |
CJ053718.1 | - | 1 - 2 | 101 | 79 | ||
U78818.2 | - | 1 - 2 | 65 | 79 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Inventoried Probe spans exons |