Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0200007002.mm.1 |
TC0200000868.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200007010.mm.1 |
NM_001145820.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200007010.mm.1 |
NM_001145820.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200007010.mm.1 |
NM_010274.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200007010.mm.1 |
NM_010274.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200007010.mm.1 |
TC0200000868.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200007001.mm.1 |
NM_001145820.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200007001.mm.1 |
NM_001145820.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200007001.mm.1 |
TC0200000868.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200007011.mm.1 |
NM_001145820.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200007011.mm.1 |
NM_001145820.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200007011.mm.1 |
NM_010274.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200007011.mm.1 |
NM_010274.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200007011.mm.1 |
TC0200000868.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200007003.mm.1 |
NM_010274.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200007003.mm.1 |
NM_010274.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200007003.mm.1 |
TC0200000868.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200007008.mm.1 |
NM_001145820.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200007008.mm.1 |
NM_001145820.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200007008.mm.1 |
NM_010274.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200007008.mm.1 |
NM_010274.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200007008.mm.1 |
TC0200000868.mm.1 |
MTA TC |