Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Grin2c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010350.2 | NP_034480.2 | 7 - 8 | 2098 | 74 | |
GenBank mRNA | AK147390.1 | - | 6 - 7 | 1826 | 74 | |
AK165770.1 | - | 4 - 5 | 953 | 74 | ||
BC137849.1 | - | 5 - 6 | 1488 | 8860868 | 74 | |
D10694.1 | - | 4 - 5 | 1325 | 74 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Ion Channel Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |